Learn More
Thomas Scientific Aspirating Pipette, 1mL, Polystyrene, Standard Tip, 278mm, Sterile, Individually Wrapped
SDP
Catalog No. 501460545
Encompass_Preferred
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes
Catalog No. 50-146-0545 Supplier Thomas Scientific Supplier No. 1209P89
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes