Learn More
Thomas Scientific Aspirating Pipette, 25mL, Polystyrene, Standard Tip, 310mm, Sterile, Individually Wrapped
SDP
Catalog No. 501460549
Encompass_Preferred
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes
Catalog No. 50-146-0549 Supplier Thomas Scientific Supplier No. 1209P93
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes