Learn More
Thomas Scientific Aspirating Pipette, 2mL, Polystyrene, Standard Tip, 278mm, Sterile, Individually Wrapped
SDP
Catalog No. 501460546
Encompass
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes
Catalog No. 50-146-0546 Supplier Thomas Scientific Supplier No. 1209P90
Sterile aspirating pipettes are completely transparent and designed for the transfer and mixing of liquids. They are also used in a wide variety of applications such as vacuum or pipettor aspirating procedures. Material: Polystyrene (PS) Transparent, no graduation marks Non-plugged Sterilized by gamma irradiation Sterility assurance level (SAL): 10 -6 Non-pyrogenic Individually wrapped in thermoformed paper/plastic peel packaging Safe alternative to glass pipettes