Learn More
McKesson General Medical ASPIRATION BLUNT CANNULA,16G
Catalog No. NC2167079
Encompass
Aspiration Blunt Cannula; 16 Gauge 3-1/2 Inch (100/BX)McKesson #: 891946Manufacturer #: 32-34Brand: Rx-Tract™Manufacturer: IMI - International Medical Industries IncCountry of Origin: United StatesApplication: Fill NeedleConnection Type: Luer LockFilter Type: Without FilterGauge: 16 GaugeHub Material: Polypropylene, PolyethyleneLength: 3-1/2 InchNeedle Material: Stainless SteelNeedle Point Style: BluntSterility: SterileSyringe Volume: Without SyringeUNSPSC Code: 42142506Usage: DisposableLatex Free Indicator: Not Made with Natural Rubber Latex
This item is not returnable. View return policy
Catalog No. NC2167079 Supplier McKesson General Medical Supplier No. 891946
Aspiration Blunt Cannula; 16 Gauge 3-1/2 Inch (100/BX)McKesson #: 891946Manufacturer #: 32-34Brand: Rx-Tract™Manufacturer: IMI - International Medical Industries IncCountry of Origin: United StatesApplication: Fill NeedleConnection Type: Luer LockFilter Type: Without FilterGauge: 16 GaugeHub Material: Polypropylene, PolyethyleneLength: 3-1/2 InchNeedle Material: Stainless SteelNeedle Point Style: BluntSterility: SterileSyringe Volume: Without SyringeUNSPSC Code: 42142506Usage: DisposableLatex Free Indicator: Not Made with Natural Rubber Latex