Learn More
Med Vet International Aspiration Kit, 2.6mm x 140cm, J0727B
Catalog No. 5000581875
Encompass_Preferred
A hollow tube with a luer attachment on one end. The flushing of saline allows a sample to be obtained (i.e., a tracheal wash via a syringe). Packaged sterile in tear-open pouches, complete with directions.
Catalog No. 50-005-81875 Supplier Med Vet International Supplier No. JOR727B
A hollow tube with a luer attachment on one end. The flushing of saline allows a sample to be obtained (i.e., a tracheal wash via a syringe). Packaged sterile in tear-open pouches, complete with directions.