Learn More
Sigma Aldrich Fine Chemicals Biosciences AspireTM Laboratory Aspira1EA
Catalog No. 502908976
Encompass
The ASPIRE Laboratory Aspirator incorporates a quiet maintenance-free pump inside a sturdy yet compact base. The vacuum is fully adjustable with a pressure gauge that displays the vacuum level allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications including removal of media from tissue culture vessels or plates and supernatants.
Catalog No. 50-290-8976 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSV00201EA
The ASPIRE Laboratory Aspirator incorporates a quiet maintenance-free pump inside a sturdy yet compact base. The vacuum is fully adjustable with a pressure gauge that displays the vacuum level allowing full control of aspirating speed. The ASPIRE is an ideal choice for a wide range of laboratory aspirating applications including removal of media from tissue culture vessels or plates and supernatants.