Learn More
Medchemexpress LLC HY-14654 500mg , Aspirin CAS:50-78-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14654500MG
Encompass_Preferred
HY-14654 500mg Aspirin CAS: 50-78-2 Aspirin (Acetylsalicylic Acid) is an orally active, potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2, with IC50 values of 5 and 210 μg/mL, respectively. Aspirin induces apoptosis. Aspirin inhibits the activation of NF-κB. Aspirin also inhibits platelet prostaglandin synthetase, and can prevent coronary artery and cerebrovascular thrombosis[4][5][6]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9361