Learn More
Apexbio Technology LLC Aspirin (Acetylsalicylic acid), 5g Cas# 50-78-2
SDP
Supplier:  Apexbio Technology LLC A40135000
Encompass_Preferred
potent and selective inhibitor of cyclooxygenase (COX) 50-78-2. MFCD00002430 Aspirin (Acetylsalicylic acid) is a potent and selective inhibitor of cyclooxygenase (COX) with a broad range of pharmacological activities including anti-inflammation and pain relief. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8583