Learn More
Medchemexpress LLC HY-B2175 500mg Medchemexpress, Aspirin Aluminum CAS:23413-80-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2175500MG
Encompass_Preferred
Medchemexpress, HY-B2175 500mg Aspirin Aluminum CAS:23413-80-1 Aspirin Aluminum is a novel intermolecular compound which can inhibit gastrointestinal mucosal disorders induced by NSAIDs (non-steroidal anti-inflammatory agents) extracted from patent WO 2010064441 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2730