Learn More
Medchemexpress LLC Aspirin-d3 | 921943-73-9 | >99.8% | 183.18 | 1 ML
SDP
Catalog No. 5000363085
Encompass_Preferred
Aspirin-d3, also known as Acetylsalicylic acid-d3 or ASA-d3, is a deuterium-labeled form of Aspirin. This orally active compound acts as a potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2. It has been shown to induce apoptosis and inhibit the activation of NF-κB, offering potential in preventing coronary artery and cerebrovascular thrombosis.
  • Deuterium labeled aspirin
  • Potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2
  • Induces apoptosis
  • Inhibits activation of NF-κB
  • May prevent coronary artery and cerebrovascular thrombosis
Catalog No. 50-003-63085 Supplier Medchemexpress LLC Supplier No. HY14654S10MM/1ML
Aspirin-d3, also known as Acetylsalicylic acid-d3 or ASA-d3, is a deuterium-labeled form of Aspirin. This orally active compound acts as a potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2. It has been shown to induce apoptosis and inhibit the activation of NF-κB, offering potential in preventing coronary artery and cerebrovascular thrombosis.
  • Deuterium labeled aspirin
  • Potent and irreversible inhibitor of cyclooxygenase COX-1 and COX-2
  • Induces apoptosis
  • Inhibits activation of NF-κB
  • May prevent coronary artery and cerebrovascular thrombosis