Learn More
Medchemexpress LLC Aspirin-d3 (Acetylsalicylic acid-d3; ASA-d3) | 921943-73-9 | 99.84% | 183.18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14654S50MG
Encompass_Preferred
Aspirin-d3 is a deuterium-labeled version of Aspirin (Acetylsalicylic acid). Aspirin is an orally active, potent, and irreversible inhibitor of cyclooxygenase COX-1 and COX-2, with IC50 values of 5 and 210 μg/mL, respectively. It is known to induce apoptosis, inhibit the activation of NF-κB, and prevent coronary artery and cerebrovascular thrombosis by inhibiting platelet prostaglandin synthetase. Deuterium labeling, as seen in Aspirin-d3, is a technique used in drug development, largely as tracers for quantitation, and can influence the pharmacokinetic and metabolic profiles of drugs. This product is intended for research use only.
  • Deuterium labeled compound
  • Stable isotope
  • Functions as a tracer in research applications
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • High purity: 99.84%
  • Available in various solid quantities and as a solution
Catalog No. 50-003-63901