Learn More
Selleck Chemical LLC Aspirin NSC 27223-50mg
SDP
Supplier:  Selleck Chemical LLC S301750MG
Encompass_Preferred
Aspirin (NSC 27223, Acetylsalicylic acid, ASA) is a salicylate, and irreversible COX1 and COX2 inhibitor, used as an analgesic to relieve minor aches and pains, as an antipyretic to reduce fever, and as an anti-inflammatory medication. Aspirin induces autophagy and stimulates mitophagy.
Catalog No. 50-238-9870