Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aspirin, USP, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18600806 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-806 50 kg Poly Bottle
18-600-805 12 kg Poly Bottle
18-600-802 100 g Poly Bottle
18-600-803 500 g Poly Bottle
18-600-804 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-806 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AS13050KGBL
Only null left
Add to Cart
Add to Cart

AS130, 50-78-2, C9H8O4

Spectrum™ Chemical Aspirin, USP is a salicylate drug that is used as an analgesic, an antipyretic, and an anti-inflammatory. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-78-2
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002430
InChI Key BSYNRYMUTXBXSQ-UHFFFAOYSA-N
IUPAC Name 2-(acetyloxy)benzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 50 kg
UN Number UN2811
Percent Purity 99.5 to 100.5%
Grade USP
Physical Form Crystalline Powder or Lumps
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.