Learn More
Aviva Systems Biology Corp ASPN Antibody - middle region (ARP42487_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42487_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ASPN. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ASPN. Alternate names: FLJ20129, PLAP1, SLRR1C, OS3, PLAP-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58244