Learn More
Medchemexpress LLC Aspochalasin D | 71968-02-0 | 95.0% | 1 MG
SDP
Catalog No. 5000342253
Encompass_Preferred
Aspochalasin D is a co-metabolite originally isolated from A. microcysticus. It exhibits antibacterial activity against Gram-positive and Gram-negative bacteria and shows cytotoxicity, via apoptosis, to Ba/F3-V12 cells, with higher cytotoxicity observed in an IL-3-free medium (IC50 = 0.49 μg/mL) compared to an IL-3-containing medium (IC50 = 1.9 μg/mL).
  • Co-metabolite isolated from A. microcysticus
  • Exhibits antibacterial activity against Gram-positive and Gram-negative bacteria
  • Shows cytotoxicity to Ba/F3-V12 cells via apoptosis
Catalog No. 50-003-42253 Supplier Medchemexpress LLC Supplier No. HY1266191MG
Aspochalasin D is a co-metabolite originally isolated from A. microcysticus. It exhibits antibacterial activity against Gram-positive and Gram-negative bacteria and shows cytotoxicity, via apoptosis, to Ba/F3-V12 cells, with higher cytotoxicity observed in an IL-3-free medium (IC50 = 0.49 μg/mL) compared to an IL-3-containing medium (IC50 = 1.9 μg/mL).
  • Co-metabolite isolated from A. microcysticus
  • Exhibits antibacterial activity against Gram-positive and Gram-negative bacteria
  • Shows cytotoxicity to Ba/F3-V12 cells via apoptosis