Learn More
AdipoGen Aspochalasin I (1 mg)
SDP
Supplier:  AdipoGen AGCN20168M001
Encompass_Preferred
Chemical. CAS: 670225-69-1. Formula: C24H35NO5. MW: 417.5. Isolated from Aspergillus sp. Cytotoxic. Melanogenesis inhibitor without cytotoxicity.
Catalog No. 50-205-3558