Learn More
Cayman Chemical Aspochalasin M, Purity: ≥95%, CAS: 1173040-34-0, 2.5 mg
SDP
Catalog No. 502934160
Encompass_Preferred
A fungal metabolite; inhibits the growth of HL-60 cells (IC50 = 20 µM)
Catalog No. 50-293-4160 Supplier Cayman Chemical Supplier No. 250872.5 MG
A fungal metabolite; inhibits the growth of HL-60 cells (IC50 = 20 µM)