Learn More
Cayman Chemical Fumagillin, Purity: ≥95%, CAS: 23110-15-8, 10 mg
SDP
Catalog No. 502951059
Encompass_Preferred
A fungal metabolite with diverse biological activities; inhibits METAP2 (IC50 = 10 nM); inhibits tube formation in a rat blood vessel organ culture assay at 10 ng/ml; inhibits E. cuniculi replication in isolated rabbit kidney cells and canine embryo cells at 5 µg/ml; decreases tumor growth and the number of metastases in a mouse model of diethylnitrosamine-induced hepatocellular carcinoma at 30 mg/kg; reduces subcutaneous and gonadal fat mass in a mouse model of high-fat diet-induced obesity
Catalog No. 50-295-1059 Supplier Cayman Chemical Supplier No. 1133210 MG
A fungal metabolite with diverse biological activities; inhibits METAP2 (IC50 = 10 nM); inhibits tube formation in a rat blood vessel organ culture assay at 10 ng/ml; inhibits E. cuniculi replication in isolated rabbit kidney cells and canine embryo cells at 5 µg/ml; decreases tumor growth and the number of metastases in a mouse model of diethylnitrosamine-induced hepatocellular carcinoma at 30 mg/kg; reduces subcutaneous and gonadal fat mass in a mouse model of high-fat diet-induced obesity