Learn More
Biomatik Corporation Asprosin Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50381
Encompass_Preferred
Asprosin Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Asprosin. The Asprosin Polyclonal Antibody has been validated for the following applications: ELISA, IHC, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 2732~2871aa. Applications: ELISA, IHC, WB. Target Synonyms: Asprosin. Accession Number: P35555. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7725