Learn More
AdipoGen Asprosin (human) Matched Pair Detection Set (1 Set)
SDP
Catalog No. 502053342
Encompass_Preferred
Matched Pair Detection Set. Detects human Asprosin in cell culture supernatant. Assay Type: Sandwich. Detection Type: Colorimetric. Sample Type: Cell Culture Supernatant. Range: 0.156 to 10ng/nl. Sensitivity: 150pg/ml. Asprosin is a new fasting-induced protein hormone that targets the liver to increase plasma glucose levels. Asprosin is the C-terminal cleavage product of the protein fibrillin-1. Asprosin is secreted from white adipose tissue and increases hepatic glucose production by using cAMP as a second messenger, leading to activation of protein kinase A in the liver. Reduction of asprosin levels protect against metabolic syndrome-associated hyperinsulinism.
Catalog No. 50-205-3342 Supplier AdipoGen Supplier No. AG46B0011KI01
Matched Pair Detection Set. Detects human Asprosin in cell culture supernatant. Assay Type: Sandwich. Detection Type: Colorimetric. Sample Type: Cell Culture Supernatant. Range: 0.156 to 10ng/nl. Sensitivity: 150pg/ml. Asprosin is a new fasting-induced protein hormone that targets the liver to increase plasma glucose levels. Asprosin is the C-terminal cleavage product of the protein fibrillin-1. Asprosin is secreted from white adipose tissue and increases hepatic glucose production by using cAMP as a second messenger, leading to activation of protein kinase A in the liver. Reduction of asprosin levels protect against metabolic syndrome-associated hyperinsulinism.