Learn More
Cayman Chemical AsprosIn PolycLNal ANTIBDY
SDP
Catalog No. 502994818
Encompass_Preferred
Immunogen: Human recombinant protein • Host: Rabbit • Species Reactivity: (+) Human asprosin • Applications: IP, WB
Catalog No. 50-299-4818 Supplier Cayman Chemical Supplier No. 23857300 UG
Immunogen: Human recombinant protein • Host: Rabbit • Species Reactivity: (+) Human asprosin • Applications: IP, WB