Learn More
Electron Microscopy Sciences Heavy Duty Vacuum Bottle w/screw cap, Material: USP Class VI medical grade resin. Cap: 83 mm., 1/EA
SDP
Catalog No. 502090871
Encompass_Preferred
Aseptic transfer caps is used with heavy duty vacuum bottles for safe, sterile transfer of reagents, media and solutions required in biotechnology, cell culture and other laboratories. Autoclavable at 121°C Material: USP Class VI medical grade resin. Cap: 83 mm .
Catalog No. 50-209-0871 Supplier Electron Microscopy Sciences Supplier No. 6426210
Aseptic transfer caps is used with heavy duty vacuum bottles for safe, sterile transfer of reagents, media and solutions required in biotechnology, cell culture and other laboratories. Autoclavable at 121°C Material: USP Class VI medical grade resin. Cap: 83 mm .