Learn More
Cayman Chemical Aspulvinone O, Purity: ≥85%, CAS: 914071-54-8, 5 mg
SDP
Catalog No. 502988668
Encompass_Preferred
A fungal metabolite; scavenges DPPH radicals in a cell-free assay (IC50 = 11.6 µM); inhibits GOT1 (Kd = 3.32 µM) and is cytotoxic to PANC-1, AsPC-1, and SW1990 pancreatic cancer cells (IC50s = 20.54-26.8 µM); reduces the OCR and induces apoptosis in SW1990 cells; reduces tumor growth in an SW1990 mouse xenograft model at 2.5 and 5 mg/kg
Catalog No. 50-298-8668 Supplier Cayman Chemical Supplier No. 299595 MG
A fungal metabolite; scavenges DPPH radicals in a cell-free assay (IC50 = 11.6 µM); inhibits GOT1 (Kd = 3.32 µM) and is cytotoxic to PANC-1, AsPC-1, and SW1990 pancreatic cancer cells (IC50s = 20.54-26.8 µM); reduces the OCR and induces apoptosis in SW1990 cells; reduces tumor growth in an SW1990 mouse xenograft model at 2.5 and 5 mg/kg