Learn More
AdipoGen Aspyrone
SDP
Supplier:  AdipoGen BVT0029M005
Encompass_Preferred
Chemical. CAS 17398-00-4. Formula C9H12O4. MW 184.2. Isolated from Aspergillus ochraceus. Antibiotic. Shows nematicidal, insecticidal, antibacterial and antifungal activity. Related to asperlactone and aspinonene.
Catalog No. 50-256-2138