Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 80150MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CRASH/1289. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HeLa cells. Fallopian Tube, Endometrium, Uterus or Cervix. Cellular Staining: Cytoplasmic and Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length ASRGL1. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 17-25kDa.Chromosome: 11q12.3. Gene Symbol: ASRGL1. Gene ID: 80150. Ensemble: ENSG00000162174. Uniprot: XP_005274363.
Catalog No. 50-188-5307