Learn More
Aviva Systems Biology Corp ASS Antibody - N-terminal region (ARP41366_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41366_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ASS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ASS. Alternate names: ASS1, CTLN1, DKFZp434P0216, ASSApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-57823