Learn More
SCANTIBODIES LABORATORY INC ASSAY DEVELOPMENT BLOCKING KIT (3KG775)
Supplier:  SCANTIBODIES LABORATORY INC 3KG775
Encompass
The 3KG775 Assay Development Blocking Kit has been designed by Scantibodies Laboratory, Inc. for those products that are currently in assay development. It contains 11 different Heterophilic Blocking Reagent (HBR) products, all of which have been carefully selected to be used as an aid in developing a reagent blocking formulation to eliminate false positive or negative interferences, and to ensure that they are useful for both active and passive blocking. This Assay Development Blocking Kit includes: HBR 1 (3KC533), HBR 3 (3KC576), HBR 6 (3KC542), HBR plus (3KC545), HBR 9 (3KC564), HBR 11 (3KC565), HBR 21 (3KC002), HBR 22 (3KC003), HBR 23 (3KC006), HBR 24 (3KC007) and HBR 26 (3KC009).
Catalog No. NC1315009