Learn More
Teknova Assay Diluent, 1000mL Sterile 2 pack.
SDP
Catalog No. 502387732
Encompass
10mM Tris-HCl,0.90% Sodium Chloride, 0.05% (v/v) Tween-20,1.00% Bovine Serum Albumin (BSA)
Catalog No. 50-238-7732 Supplier Teknova Supplier No. 2A30502PK
10mM Tris-HCl,0.90% Sodium Chloride, 0.05% (v/v) Tween-20,1.00% Bovine Serum Albumin (BSA)