Learn More
Surmodics Inc Assay Diluent (Protein-Free), 50mL
SDP
Catalog No. NC0450512
Encompass_Preferred
The Surmodics™ Assay Diluent (Protein-Free) is a universal blocking agent that utilizes synthetic components to reduce matrix interference within immunoassay applications. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies (HAMA and rheumatoid factor).
Catalog No. NC0450512 Supplier Surmodics Inc Supplier No. SM010050
The Surmodics™ Assay Diluent (Protein-Free) is a universal blocking agent that utilizes synthetic components to reduce matrix interference within immunoassay applications. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies (HAMA and rheumatoid factor).