Learn More
Surmodics Inc Assay Diluent (Protein-Free), 1L
SDP
Catalog No. NC0734917
Encompass_Preferred
The Surmodics™ Assay Diluent (Protein-Free) is a universal blocking agent that utilizes synthetic components to reduce matrix interference within immunoassay applications. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies (HAMA and rheumatoid factor).
Catalog No. NC0734917 Supplier Surmodics Inc Supplier No. SM011000
The Surmodics™ Assay Diluent (Protein-Free) is a universal blocking agent that utilizes synthetic components to reduce matrix interference within immunoassay applications. Increased signal-to-noise ratios are produced when using the product with samples containing heterophilic antibodies (HAMA and rheumatoid factor).