Learn More
Genscript Corporation Assay Dilution Buffer
Catalog No. 5000058348
Encompass
Assay Dilution Buffer is Ready-to-use and designed to dilute high concentration samples or standards and minimize sample matrix interferences
Catalog No. 50-000-58348 Supplier Genscript Corporation Supplier No. B00062
Assay Dilution Buffer is Ready-to-use and designed to dilute high concentration samples or standards and minimize sample matrix interferences