Learn More
Molecular Devices Molecular Devices FLIPR Calcium 6 Assay Kit, 10-Plate Explorer
Catalog No. 5000014342
Encompass_Preferred
The Molecular Devices FLIPR Calcium 6 Explorer Kit provides reagents for fluorescent intracellular calcium flux assays optimized for FLIPR-compatible high-throughput readers. The kit supplies enough reagent for ten assay plates and is engineered to deliver a high signal-to-background window for low-signal targets such as endogenous, primary, or stem cell assays.
  • Explorer kit format supplies reagents for 10 assay plates.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Includes 10 vials of Component A and 1 bottle of dilution/assay buffer (Component B).
  • Optimized calcium indicator with high quantum yield for strong signal-to-background.
  • Suitable for FLIPR-compatible plate readers and high-throughput calcium flux screening.
Catalog No. 50-000-14342 Supplier Molecular Devices Supplier No. R8190
The Molecular Devices FLIPR Calcium 6 Explorer Kit provides reagents for fluorescent intracellular calcium flux assays optimized for FLIPR-compatible high-throughput readers. The kit supplies enough reagent for ten assay plates and is engineered to deliver a high signal-to-background window for low-signal targets such as endogenous, primary, or stem cell assays.
  • Explorer kit format supplies reagents for 10 assay plates.
  • Compatible with 96-well, 384-well, and 1536-well plate formats.
  • Includes 10 vials of Component A and 1 bottle of dilution/assay buffer (Component B).
  • Optimized calcium indicator with high quantum yield for strong signal-to-background.
  • Suitable for FLIPR-compatible plate readers and high-throughput calcium flux screening.