Learn More
Epigentek Inc EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3K27) (48 assays)
SDP
Catalog No. NC0013496
Encompass_Preferred
The EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K27) is a convenient set of tools that allows the experimenter to measure the activity or inhibition of individual histone methyltransferase that specifically target histone H3 at lysine 27 (H3-K27). The kit is ready-to-use and provides all the essential components needed to carry out a successful HMT activity/inhibition experiment without the need for radioactivity or any special equipment.
Catalog No. NC0013496 Supplier Epigentek Inc Supplier No. P300548
The EpiQuik Histone Methyltransferase Activity/Inhibition Assay Kit (H3-K27) is a convenient set of tools that allows the experimenter to measure the activity or inhibition of individual histone methyltransferase that specifically target histone H3 at lysine 27 (H3-K27). The kit is ready-to-use and provides all the essential components needed to carry out a successful HMT activity/inhibition experiment without the need for radioactivity or any special equipment.