Learn More
Waters Corp Waters Tubing Assembly, MP35N, 0.062 in. x 0.004 in. x 25 in., Low Pressure
Catalog No. NC2199677 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement tubing assembly is designed for use in Bio Systems with a 30 cm column heater. It is constructed from MP35N material and features low pressure fittings.
  • Constructed from MP35N material.
  • Provided with low pressure fittings.
  • Suitable for Bio Systems.
  • Compatible with a 30 cm column heater.
This item is not returnable. View return policy
Catalog No. NC2199677 Supplier Waters Corp Supplier No. 700011810
This replacement tubing assembly is designed for use in Bio Systems with a 30 cm column heater. It is constructed from MP35N material and features low pressure fittings.
  • Constructed from MP35N material.
  • Provided with low pressure fittings.
  • Suitable for Bio Systems.
  • Compatible with a 30 cm column heater.