Learn More
MyBioSource NA/K ATPase
SDP
Catalog No. 501689750
Encompass_Preferred
The MyBioSource NA/K ATPase ELISA Kit quantitatively detects NA/K ATPase in various biological samples using a sandwich enzyme immunoassay. It involves binding, washing, and colorimetric detection, with a range of 0.312-20 ng/ml and sensitivity of 0.188 ng/ml.
Catalog No. 50-168-9750 Supplier MyBioSource Supplier No. MBS8243226100ASSAYS
The MyBioSource NA/K ATPase ELISA Kit quantitatively detects NA/K ATPase in various biological samples using a sandwich enzyme immunoassay. It involves binding, washing, and colorimetric detection, with a range of 0.312-20 ng/ml and sensitivity of 0.188 ng/ml.