Learn More
Molecular Devices Molecular Devices Donkey Anti-Goat Secondary Antibody, Europium-Labeled, 60 μg
Catalog No. 5000014453
Encompass_Preferred
The Molecular Devices ScanLater Eu-Labeled Donkey Anti-Goat IgG is a Europium-labeled secondary antibody supplied as a 60 μg vial for time-resolved fluorescence detection of goat primary antibodies in western blot applications. It is formulated and validated for use with TRF-based microplate reader detection workflows and intended for research use only.
  • Europium-labeled secondary antibody for time-resolved fluorescence detection.
  • Supplied as a 60 μg vial.
  • Optimized for western blot detection with TRF microplate readers.
  • Suitable for detection of goat primary antibodies in protein transfer membranes.
  • For research use only; not for diagnostic procedures.
Catalog No. 50-000-14453 Supplier Molecular Devices Supplier No. R8226
The Molecular Devices ScanLater Eu-Labeled Donkey Anti-Goat IgG is a Europium-labeled secondary antibody supplied as a 60 μg vial for time-resolved fluorescence detection of goat primary antibodies in western blot applications. It is formulated and validated for use with TRF-based microplate reader detection workflows and intended for research use only.
  • Europium-labeled secondary antibody for time-resolved fluorescence detection.
  • Supplied as a 60 μg vial.
  • Optimized for western blot detection with TRF microplate readers.
  • Suitable for detection of goat primary antibodies in protein transfer membranes.
  • For research use only; not for diagnostic procedures.