Learn More
BPS Bioscience Inc Assay Medium 1B | 100mL
SDP
Catalog No. NC1553313
Encompass
This liquid Assay Medium is optimized for use in cellular assays in select BPS Bioscience’s cell lines, including: TGF/SMAD Signaling Pathway SBE Reporter – HEK293 Cell Line #60653, AP-1 Reporter – HEK293 Recombinant Cell Line (JNK signaling pathway) #60405
Catalog No. NC1553313 Supplier BPS Bioscience Inc Supplier No. 796171
This liquid Assay Medium is optimized for use in cellular assays in select BPS Bioscience’s cell lines, including: TGF/SMAD Signaling Pathway SBE Reporter – HEK293 Cell Line #60653, AP-1 Reporter – HEK293 Recombinant Cell Line (JNK signaling pathway) #60405