Learn More
Molecular Devices Molecular Devices IMAP FP Screening Kit, Progressive Binding, Fluorescence Polarization
Catalog No. 5000014388
Encompass_Preferred
The IMAP FP Screening Express Kit is a fluorescence polarization reagent kit for high-throughput enzyme screening and assay development. It provides IMAP binding reagent and binding buffers configured with a Progressive Binding System to optimize homogeneous FP assays for kinases, phosphatases, and phosphodiesterases in a 384-well format.
  • Homogeneous fluorescence polarization readout for enzyme activity
  • Progressive Binding System for assay condition optimization
  • Includes IMAP binding reagent and binding buffers
  • Sufficient reagents for approximately 8,000 data points in 384-well plates
  • Labeled substrates and additional substrates sold separately
  • Suitable for kinase, phosphatase, and phosphodiesterase assays
Catalog No. 50-000-14388 Supplier Molecular Devices Supplier No. R8127
Add to Cart
Add to Cart
Request a Quote
The IMAP FP Screening Express Kit is a fluorescence polarization reagent kit for high-throughput enzyme screening and assay development. It provides IMAP binding reagent and binding buffers configured with a Progressive Binding System to optimize homogeneous FP assays for kinases, phosphatases, and phosphodiesterases in a 384-well format.
  • Homogeneous fluorescence polarization readout for enzyme activity
  • Progressive Binding System for assay condition optimization
  • Includes IMAP binding reagent and binding buffers
  • Sufficient reagents for approximately 8,000 data points in 384-well plates
  • Labeled substrates and additional substrates sold separately
  • Suitable for kinase, phosphatase, and phosphodiesterase assays