Learn More
Molecular Devices Molecular Devices CatchPoint cAMP Assay Kit, Fluorescent, 384-Well, Explorer
Catalog No. 5000014457
Encompass_Preferred
The CatchPoint cAMP Fluorescent Assay Kit provides a sensitive, quantitative fluorescence-based competitive immunoassay for measuring cyclic AMP (cAMP) in high-throughput 384-well workflows. The Explorer kit includes ready-to-use plates and reagents optimized for low cAMP levels and automation-compatible assays.
  • Quantitative competitive fluorescent immunoassay for cAMP
  • 384-well black well/clear-bottom plates, pre-coated and pre-blocked
  • Includes two 384-well assay plates and required reagents and buffers
  • Stoplight Red substrate provides a stable fluorescent readout
  • Flexible read times from about 10 minutes up to 24 hours after substrate addition
  • Compatible with high-throughput screening and automation
Catalog No. 50-000-14457 Supplier Molecular Devices Supplier No. R8044
Add to Cart
Add to Cart
The CatchPoint cAMP Fluorescent Assay Kit provides a sensitive, quantitative fluorescence-based competitive immunoassay for measuring cyclic AMP (cAMP) in high-throughput 384-well workflows. The Explorer kit includes ready-to-use plates and reagents optimized for low cAMP levels and automation-compatible assays.
  • Quantitative competitive fluorescent immunoassay for cAMP
  • 384-well black well/clear-bottom plates, pre-coated and pre-blocked
  • Includes two 384-well assay plates and required reagents and buffers
  • Stoplight Red substrate provides a stable fluorescent readout
  • Flexible read times from about 10 minutes up to 24 hours after substrate addition
  • Compatible with high-throughput screening and automation