Learn More
MTC Bio Assay Plates (PS) 384 Square Well Flat-Bottom Natural 14 41mm Height 110uL With Lid Sterile Individually Wrapped 1 Plate/Pack 40 Packs/Case
Catalog No. 5000460195
Encompass_Preferred
Assay Plates (PS) 384 Square Well Flat-Bottom Natural 14 41mm Height 110uL With Lid Sterile Individually Wrapped 1 Plate/Pack 40 Packs/Case
Catalog No. 50-004-60195 Supplier MTC Bio Supplier No. DP110F384NSIWL
Add to Cart
Add to Cart
Assay Plates (PS) 384 Square Well Flat-Bottom Natural 14 41mm Height 110uL With Lid Sterile Individually Wrapped 1 Plate/Pack 40 Packs/Case