Learn More
MTC Bio Assay Plates (PS) 96 Round Well Flat-Bottom Natural 14 10mm Height 340uL Non-Sterile 10 Plates/Pack 10 Packs/Case
Catalog No. 5000460176
Encompass_Preferred
Assay Plates (PS) 96 Round Well Flat-Bottom Natural 14 10mm Height 340uL Non-Sterile 10 Plates/Pack 10 Packs/Case
Catalog No. 50-004-60176 Supplier MTC Bio Supplier No. DP35F96N
Assay Plates (PS) 96 Round Well Flat-Bottom Natural 14 10mm Height 340uL Non-Sterile 10 Plates/Pack 10 Packs/Case