Learn More
Assay Technology Assay Technology Ammonia Monitor, Personal Exposure, Ion Chromatography, STEL or TWA Monitoring
SDP
Catalog No. NC3978722
Encompass_Preferred
The 584 Ammonia Monitor is designed for measuring personal exposure to ammonia vapor. Ammonia is collected on fiberglass treated with sulfuric acid, then desorbed with deionized water and analyzed as ammonium ion using Ion Chromatography.
  • Measures personal exposure to ammonia vapor.
  • Collects ammonia on fiberglass treated with sulfuric acid.
  • Analyzes ammonia as ammonium ion using ion chromatography.
  • Suitable for 15-minute STEL sampling.
  • Suitable for 8-hour TWA sampling.
  • Associated analytical method: OSHA 188 Modified.
  • Requires refrigeration before sampling for maximum shelf life.
  • Store and ship at room temperature after sampling; samples to arrive at laboratory within 14 days.
Catalog No. NC3978722 Supplier Assay Technology Supplier No. X584AT
The 584 Ammonia Monitor is designed for measuring personal exposure to ammonia vapor. Ammonia is collected on fiberglass treated with sulfuric acid, then desorbed with deionized water and analyzed as ammonium ion using Ion Chromatography.
  • Measures personal exposure to ammonia vapor.
  • Collects ammonia on fiberglass treated with sulfuric acid.
  • Analyzes ammonia as ammonium ion using ion chromatography.
  • Suitable for 15-minute STEL sampling.
  • Suitable for 8-hour TWA sampling.
  • Associated analytical method: OSHA 188 Modified.
  • Requires refrigeration before sampling for maximum shelf life.
  • Store and ship at room temperature after sampling; samples to arrive at laboratory within 14 days.