Learn More
Eurofins Discoverx AssayComplete Freezing Reagent F3, 2 x 50. (92-5103FR)
Catalog No. NC1320667
Encompass
AssayComplete Freezing Reagent F3 is an optimized freezing reagent for cryopreservation of Eurofins DiscoverX cell lines. This reagent ensures optimal cell viability and assay performance upon thawing. This product is provided as 2 bottles of 50 mL reagent each. Please refer to cell line specific datasheet to determine the recommended freezing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1320667 Supplier Eurofins Discoverx Supplier No. 925103FR
AssayComplete Freezing Reagent F3 is an optimized freezing reagent for cryopreservation of Eurofins DiscoverX cell lines. This reagent ensures optimal cell viability and assay performance upon thawing. This product is provided as 2 bottles of 50 mL reagent each. Please refer to cell line specific datasheet to determine the recommended freezing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.