Learn More
Eurofins Discoverx AssayComplete Hygromycin B, 20. (92-0029)
Catalog No. NC1302347
Encompass
AssayComplete Hygromycin B is ready-to-use, sterile-filtered hygromycin B antibiotic shipped as a 50 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 20 mL bottle of liquid at 50 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1302347 Supplier Eurofins Discoverx Supplier No. 920029
AssayComplete Hygromycin B is ready-to-use, sterile-filtered hygromycin B antibiotic shipped as a 50 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 20 mL bottle of liquid at 50 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.