Learn More
Eurofins Discoverx AssayComplete G418, 20. (92-0030)
Catalog No. NC1169049
Encompass
AssayComplete G418 is ready-to-use, sterile-filtered G418 antibiotic shipped as a 50 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 20 mL bottle of liquid at 50 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1169049 Supplier Eurofins Discoverx Supplier No. 920030
AssayComplete G418 is ready-to-use, sterile-filtered G418 antibiotic shipped as a 50 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 20 mL bottle of liquid at 50 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.