Learn More
Eurofins Discoverx AssayComplete Protein Dilution Buffer, 2 x 250. (92-0023L)
Catalog No. NC1507484
Encompass
AssayComplete Protein Dilution Buffer is a specially formulated dilution buffer designed for use in reconstituting protein agonists and diluting other ligands (antibodies, small molecules, etc.) to the proper stock concentration for use in Eurofins DiscoverX cell-based assays. This product is provided as 2 bottles of 50 mL buffer each. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1507484 Supplier Eurofins Discoverx Supplier No. 920023L
AssayComplete Protein Dilution Buffer is a specially formulated dilution buffer designed for use in reconstituting protein agonists and diluting other ligands (antibodies, small molecules, etc.) to the proper stock concentration for use in Eurofins DiscoverX cell-based assays. This product is provided as 2 bottles of 50 mL buffer each. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.