Learn More
Eurofins Discoverx AssayComplete Puromycin, 1. (92-0028)
Catalog No. NC1278826
Encompass
AssayComplete Puromycin is ready-to-use, sterile-filtered puromycin antibiotic shipped as a 10 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 1 mL bottle of liquid at 10 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1278826 Supplier Eurofins Discoverx Supplier No. 920028
AssayComplete Puromycin is ready-to-use, sterile-filtered puromycin antibiotic shipped as a 10 mg/mL liquid stock. This antibiotic is intended to be used as a selection agent in culture of Eurofins DiscoverX cell-based assays. This product is provided as a 1 mL bottle of liquid at 10 mg/mL. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.