Learn More
Eurofins Discoverx AssayComplete Thawing Reagent T6, 100. (92-4106TR)
Catalog No. NC1177745
Encompass
AssayComplete Thawing Reagent T6 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1177745 Supplier Eurofins Discoverx Supplier No. 924106TR
AssayComplete Thawing Reagent T6 is an optimized thawing reagent for revival and recovery of Eurofins DiscoverX cell lines. This reagent ensures optimal post-thaw viability, recovery and assay performance upon thawing. This product is provided as a bottle of 100 mL. Please refer to cell line specific datasheet to determine the recommended thawing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.