Learn More
Eurofins Discoverx AssayComplete Freezing Reagent F5, 2 x 50. (92-5105FR)
Catalog No. NC1177746
Encompass
AssayComplete Freezing Reagent F5 is an optimized freezing reagent for cryopreservation of Eurofins DiscoverX cell lines. This reagent ensures optimal cell viability and assay performance upon thawing. This product is provided as 2 bottles of 50 mL reagent each. Please refer to cell line specific datasheet to determine the recommended freezing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC1177746 Supplier Eurofins Discoverx Supplier No. 925105FR
AssayComplete Freezing Reagent F5 is an optimized freezing reagent for cryopreservation of Eurofins DiscoverX cell lines. This reagent ensures optimal cell viability and assay performance upon thawing. This product is provided as 2 bottles of 50 mL reagent each. Please refer to cell line specific datasheet to determine the recommended freezing reagent. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.