Learn More
Randox ASSAYD CHEM PREM PLUS CONT III
SDP
Catalog No. NC9666244
Encompass_Preferred
Lyophilized assayed chemistry control with method & instrument specific target values and ranges for 70 analytes. Reconstituted Stability of 4 weeks at -20C or 7 days at +2-8C.
Catalog No. NC9666244 Supplier Randox Supplier No. HE1532
Lyophilized assayed chemistry control with method & instrument specific target values and ranges for 70 analytes. Reconstituted Stability of 4 weeks at -20C or 7 days at +2-8C.